MilliporeSigma
  • A mitochondrial NADPH-cholesterol axis regulates extracellular vesicle biogenesis to support hematopoietic stem cell fate.
A mitochondrial NADPH-cholesterol axis regulates extracellular vesicle biogenesis to support hematopoietic stem cell fate.
Cell stem cell (2024-03-09)
Massimo Bonora, Claudia Morganti, Nick van Gastel, Kyoko Ito, Enrica Calura, Ilaria Zanolla, Letizia Ferroni, Yang Zhang, Yookyung Jung, Gabriele Sales, Paolo Martini, Takahisa Nakamura, Francesco Massimo Lasorsa, Toren Finkel, Charles P Lin, Barbara Zavan, Paolo Pinton, Irene Georgakoudi, Chiara Romualdi, David T Scadden, Keisuke Ito
ABSTRACT
Mitochondrial fatty acid oxidation (FAO) is essential for hematopoietic stem cell (HSC) self-renewal; however, the mechanism by which mitochondrial metabolism controls HSC fate remains unknown. Here, we show that within the hematopoietic lineage, HSCs have the largest mitochondrial NADPH pools, which are required for proper HSC cell fate and homeostasis. Bioinformatic analysis of the HSC transcriptome, biochemical assays, and genetic inactivation of FAO all indicate that FAO-generated NADPH fuels cholesterol synthesis in HSCs. Interference with FAO disturbs the segregation of mitochondrial NADPH toward corresponding daughter cells upon single HSC division. Importantly, we have found that the FAO-NADPH-cholesterol axis drives extracellular vesicle (EV) biogenesis and release in HSCs, while inhibition of EV signaling impairs HSC self-renewal. These data reveal the existence of a mitochondrial NADPH-cholesterol axis for EV biogenesis that is required for hematopoietic homeostasis and highlight the non-stochastic nature of HSC fate determination.
Sigma-Aldrich
(+)-Etomoxir sodium salt hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-Farnesyl Rabbit pAb, liquid, Calbiochem®
Sigma-Aldrich
BMS-303141, ≥98% (HPLC)
Sigma-Aldrich
GW4869, ≥90% (NMR)