MilliporeSigma
  • Cardioprotective properties of OMT-28, a synthetic analog of omega-3 epoxyeicosanoids.
Cardioprotective properties of OMT-28, a synthetic analog of omega-3 epoxyeicosanoids.
The Journal of biological chemistry (2024-05-17)
Joshua Kranrod, Anne Konkel, Robert Valencia, Ahmed M Darwesh, Robert Fischer, Wolf-Hagen Schunck, John M Seubert
ABSTRACT
OMT-28 is a metabolically robust small molecule developed to mimic the structure and function of omega-3 epoxyeicosanoids. However, it remained unknown to what extent OMT-28 also shares the cardioprotective and anti-inflammatory properties of its natural counterparts. To address this question, we analyzed the ability of OMT-28 to ameliorate hypoxia/reoxygenation (HR)-injury and lipopolysaccharide (LPS)-induced endotoxemia in cultured cardiomyocytes. Moreover, we investigated the potential of OMT-28 to limit functional damage and inflammasome activation in isolated perfused mouse hearts subjected to ischemia/reperfusion (IR) injury. In the HR model, OMT-28 (1 μM) treatment largely preserved cell viability (about 75 versus 40% with the vehicle) and mitochondrial function as indicated by the maintenance of NAD+/NADH-, ADP/ATP-, and respiratory control ratios. Moreover, OMT-28 blocked the HR-induced production of mitochondrial reactive oxygen species. Pharmacological inhibition experiments suggested that Gαi, PI3K, PPARα, and Sirt1 are essential components of the OMT-28-mediated pro-survival pathway. Counteracting inflammatory injury of cardiomyocytes, OMT-28 (1 μM) reduced LPS-induced increases in TNFα protein (by about 85% versus vehicle) and NF-κB DNA binding (by about 70% versus vehicle). In the ex vivo model, OMT-28 improved post-IR myocardial function recovery to reach about 40% of the baseline value compared to less than 20% with the vehicle. Furthermore, OMT-28 (1 μM) limited IR-induced NLRP3 inflammasome activation similarly to a direct NLRP3 inhibitor (MCC950). Overall, this study demonstrates that OMT-28 possesses potent cardio-protective and anti-inflammatory properties supporting the hypothesis that extending the bioavailability of omega-3 epoxyeicosanoids may improve their prospects as therapeutic agents.
Sigma-Aldrich
Wortmannin, Wortmannin, CAS 19545-26-7, is a cell-permeable, potent, selective, and irreversible inhibitor of PI3-Kinase (IC₅₀ = 5 nM). Does not affect any upstream signaling events.
Sigma-Aldrich
EX-527, ≥98% (HPLC)
Sigma-Aldrich
L-(–)-Epinephrine-(+)-bitartrate, α,β-Adrenergic receptor agonist.
Sigma-Aldrich
CP-456773 sodium salt, ≥98% (HPLC)