MilliporeSigma
Regulation of floral organ abscission in Arabidopsis thaliana.
Proceedings of the National Academy of Sciences of the United States of America (2008-09-24)
Sung Ki Cho, Clayton T Larue, David Chevalier, Huachun Wang, Tsung-Luo Jinn, Shuqun Zhang, John C Walker
ABSTRACT
Abscission is a developmental program that results in the active shedding of infected or nonfunctional organs from a plant body. Here, we establish a signaling pathway that controls abscission in Arabidopsis thaliana from ligand, to receptors, to downstream effectors. Loss of function mutations in Inflorescence Deficient in Abscission (IDA), which encodes a predicted secreted small protein, the receptor-like protein kinases HAESA (HAE) and HAESA-like 2 (HSL2), the Mitogen-Activated Protein Kinase Kinase 4 (MKK4) and MKK5, and a dominant-negative form of Mitogen-Activated Protein Kinase 6 (MPK6) in a mpk3 mutant background all have abscission-defective phenotypes. Conversely, expression of constitutively active MKKs rescues the abscission-defective phenotype of hae hsl2 and ida plants. Additionally, in hae hsl2 and ida plants, MAP kinase activity is reduced in the receptacle, the part of the stem that holds the floral organs. Plants overexpressing IDA in a hae hsl2 background have abscission defects, indicating HAE and HSL2 are epistatic to IDA. Taken together, these results suggest that the sequential action of IDA, HAE and HSL2, and a MAP kinase cascade regulates the programmed separation of cells in the abscission zone.
Sigma-Aldrich
Hygromycin B from Streptomyces hygroscopicus, suitable for plant cell culture, BioReagent, ≥60% (HPLC), lyophilized powder
Sigma-Aldrich
Hygromycin B from Streptomyces hygroscopicus, lyophilized powder
Sigma-Aldrich
Hygromycin B from Streptomyces hygroscopicus, powder, BioReagent, suitable for cell culture, suitable for insect cell culture