MilliporeSigma
  • Translocation of nuclear chromatin distribution to the periphery reflects dephosphorylated threonine-821/826 of the retinoblastoma protein (pRb) in T24 cells treated with Bacillus Calmette-Guérin.
Translocation of nuclear chromatin distribution to the periphery reflects dephosphorylated threonine-821/826 of the retinoblastoma protein (pRb) in T24 cells treated with Bacillus Calmette-Guérin.
Cytotechnology (2023-01-31)
Toshitaka Uehara, Sumiko Watanabe, Shota Yamaguchi, Natsuki Eguchi, Norie Sakamoto, Yoshinao Oda, Hidetaka Arimura, Tsunehisa Kaku, Yoshihiro Ohishi, Shinichi Mizuno
ABSTRACT
The standard treatment for non-muscle-invasive bladder cancer is intravesical Bacillus Calmette-Guérin (BCG) therapy, which is considered the only intravesical therapy that reduces the risk of progression to muscle-invasive cancer. BCG unresponsiveness, in which intravesical BCG therapy is ineffective, has become a problem. It is thus important to evaluate the effectiveness of BCG treatment for patients as soon as possible in order to identify the optimal therapy. Urine cytology is a noninvasive, easy, and cost-effective method that has been used during BCG treatment, but primarily only to determine benign or malignant status; findings concerning the efficacy of BCG treatment based on urine cytology have not been reported. We investigated the relationship between BCG exposure and nuclear an important criterion in urine cytology, i.e., nuclear chromatin patterns. We used three types of cultured cells to evaluate nuclear chromatin patterns and the cell cycle, and we used T24 cells to evaluate the phosphorylation of retinoblastoma protein (pRb) in six-times of BCG exposures. The results revealed that after the second BCG exposure, (i) nuclear chromatin is distributed predominantly at the nuclear periphery and (ii) the dephosphorylation of threonine-821/826 in pRb occurs. This is the first report of a dynamic change in the nuclear chromatin pattern induced by exposure to BCG. Molecular findings also suggested a relationship between this phenomenon and cell-cycle proteins. Although these results are preliminary, they contribute to our understanding of the cytomorphological changes that occur with BCG exposure.
Sigma-Aldrich
Anti-Akt1/PKBα Antibody, clone AW24, rabbit monoclonal, clone AW24, Upstate®, from rabbit