MilliporeSigma
  • A plant NLR receptor employs ABA central regulator PP2C-SnRK2 to activate antiviral immunity.
A plant NLR receptor employs ABA central regulator PP2C-SnRK2 to activate antiviral immunity.
Nature communications (2024-04-14)
Shen Huang, Chunli Wang, Zixuan Ding, Yaqian Zhao, Jing Dai, Jia Li, Haining Huang, Tongkai Wang, Min Zhu, Mingfeng Feng, Yinghua Ji, Zhongkai Zhang, Xiaorong Tao
ABSTRACT
Defence against pathogens relies on intracellular nucleotide-binding, leucine-rich repeat immune receptors (NLRs) in plants. Hormone signaling including abscisic acid (ABA) pathways are activated by NLRs and play pivotal roles in defence against different pathogens. However, little is known about how hormone signaling pathways are activated by plant immune receptors. Here, we report that a plant NLR Sw-5b mimics the behavior of the ABA receptor and directly employs the ABA central regulator PP2C-SnRK2 complex to activate an ABA-dependent defence against viral pathogens. PP2C4 interacts with and constitutively inhibits SnRK2.3/2.4. Behaving in a similar manner as the ABA receptor, pathogen effector ligand recognition triggers the conformational change of Sw-5b NLR that enables binding to PP2C4 via the NB domain. This receptor-PP2C4 binding interferes with the interaction between PP2C4 and SnRK2.3/2.4, thereby releasing SnRK2.3/2.4 from PP2C4 inhibition to activate an ABA-specific antiviral immunity. These findings provide important insights into the activation of hormone signaling pathways by plant immune receptors.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
MONOCLONAL ANTI-GST TAG antibody produced in mouse, clone 9AT106, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-GFP antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution