MilliporeSigma
  • Selectivity of TMC207 towards mycobacterial ATP synthase compared with that towards the eukaryotic homologue.
Selectivity of TMC207 towards mycobacterial ATP synthase compared with that towards the eukaryotic homologue.
Antimicrobial agents and chemotherapy (2008-12-17)
Anna C Haagsma, Rooda Abdillahi-Ibrahim, Marijke J Wagner, Klaas Krab, Karen Vergauwen, Jerome Guillemont, Koen Andries, Holger Lill, Anil Koul, Dirk Bald
ABSTRACT
The diarylquinoline TMC207 kills Mycobacterium tuberculosis by specifically inhibiting ATP synthase. We show here that human mitochondrial ATP synthase (50% inhibitory concentration [IC(50)] of >200 microM) displayed more than 20,000-fold lower sensitivity for TMC207 compared to that of mycobacterial ATP synthase (IC(50) of 10 nM). Also, oxygen consumption in mouse liver and bovine heart mitochondria showed very low sensitivity for TMC207. These results suggest that TMC207 may not elicit ATP synthesis-related toxicity in mammalian cells. ATP synthase, although highly conserved between prokaryotes and eukaryotes, may still qualify as an attractive antibiotic target.
Sigma-Aldrich
TMC207, ≥98% (HPLC)