MilliporeSigma
  • The relation between urinary incontinence and steroid hormone levels in perimenopausal women. A report from the Women's Health in the Lund Area (WHILA) study.
The relation between urinary incontinence and steroid hormone levels in perimenopausal women. A report from the Women's Health in the Lund Area (WHILA) study.
Acta obstetricia et gynecologica Scandinavica (2009-07-07)
Pia M Teleman, Jan Persson, Anders Mattiasson, Goran Samsioe
ABSTRACT
To outline possible associations between urinary incontinence (UI) and serum levels of steroid hormones in middle-aged women. Community-based observational study. All women aged 50-59 living in the Lund area by December 1995 were invited to a screening procedure. Sixty-four percent (n = 6,917) attended the screening that included physical and laboratory examinations and questionnaires. Serum levels of cortisol, testosterone, androstendione, SHBG (sex hormone-binding globulin), and estradiol were analyzed and the 2,221 (32%) women who reported urinary leakage causing a social or hygienic problem were compared to those who denied incontinence. Possible differences in serum levels of steroid hormones in continent and incontinent women. There were no significant differences between continent and incontinent women regarding serum levels of cortisol, testosterone, androstendione, or testosterone + androstendione combined. Serum estradiol adjusted for body mass index, parity, smoking, and hysterectomy was significantly higher in incontinent women (87.1 +/- 138.4 pmol/l vs. 78.0 +/- 118.5 pmol/l, p = 0.005), whereas the ratio estradiol/SHBG was not. These differences persisted when the group of women not on hormonal treatment was analyzed. UI in middle-aged women seems related to higher serum estradiol levels. This corroborates with studies showing a higher incidence and/or prevalence of UI in women on hormone therapy. No association between UI and serum levels of cortisol, testosterone, or androstendione was found.
Sigma-Aldrich
Differentiation Solution, for the differentiation of regressive hematoxylin stains