MilliporeSigma
  • Delineating the early dissemination mechanisms of acral melanoma by integrating single-cell and spatial transcriptomic analyses.
Delineating the early dissemination mechanisms of acral melanoma by integrating single-cell and spatial transcriptomic analyses.
Nature communications (2023-12-09)
Chuanyuan Wei, Wei Sun, Kangjie Shen, Jingqin Zhong, Wanlin Liu, Zixu Gao, Yu Xu, Lu Wang, Tu Hu, Ming Ren, Yinlam Li, Yu Zhu, Shaoluan Zheng, Ming Zhu, Rongkui Luo, Yanwen Yang, Yingyong Hou, Fazhi Qi, Yuhong Zhou, Yong Chen, Jianying Gu
ABSTRACT
Acral melanoma (AM) is a rare subtype of melanoma characterized by a high incidence of lymph node (LN) metastasis, a critical factor in tumor dissemination and therapeutic decision-making. Here, we employ single-cell and spatial transcriptomic analyses to investigate the dynamic evolution of early AM dissemination. Our findings reveal substantial inter- and intra-tumor heterogeneity in AM, alongside a highly immunosuppressive tumor microenvironment and complex intercellular communication networks, particularly in patients with LN metastasis. Notably, we identify a strong association between MYC+ Melanoma (MYC+MEL) and FGFBP2+NKT cells with LN metastasis. Furthermore, we demonstrate that LN metastasis requires a metabolic shift towards fatty acid oxidation (FAO) induced by MITF in MYC+MEL cells. Etomoxir, a clinically approved FAO inhibitor, can effectively suppress MITF-mediated LN metastasis. This comprehensive dataset enhances our understanding of LN metastasis in AM, and provides insights into the potential therapeutic targeting for the management of early AM dissemination.
Sigma-Aldrich
Anti-FGFBP2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution