MilliporeSigma
  • HIV-1 Establishes a Sanctuary Site in the Testis by Permeating the BTB Through Changes in Cytoskeletal Organization.
HIV-1 Establishes a Sanctuary Site in the Testis by Permeating the BTB Through Changes in Cytoskeletal Organization.
Endocrinology (2021-08-04)
Siwen Wu, Ines Frank, Nina Derby, Elena Martinelli, C Yan Cheng
ABSTRACT
Studies suggest that HIV-1 invades the testis through initial permeation of the blood-testis barrier (BTB). The selectivity of the BTB to antiretroviral drugs makes this site a sanctuary for the virus. Little is known about how HIV-1 crosses the BTB and invades the testis. Herein, we used 2 approaches to examine the underlying mechanism(s) by which HIV-1 permeates the BTB and gains entry into the seminiferous epithelium. First, we examined if recombinant Tat protein was capable of perturbing the BTB and making the barrier leaky, using the primary rat Sertoli cell in vitro model that mimics the BTB in vivo. Second, we used HIV-1-infected Sup-T1 cells to investigate the activity of HIV-1 infection on cocultured Sertoli cells. Using both approaches, we found that the Sertoli cell tight junction permeability barrier was considerably perturbed and that HIV-1 effectively permeates the BTB by inducing actin-, microtubule-, vimentin-, and septin-based cytoskeletal changes in Sertoli cells. These studies suggest that HIV-1 directly perturbs BTB function, potentially through the activity of the Tat protein.
Sigma-Aldrich
Anti-ARP3 antibody, Mouse monoclonal, clone FMS338, purified from hybridoma cell culture
Sigma-Aldrich
Anti-alpha Tubulin Antibody, tyrosinated, serum, from rabbit