MilliporeSigma
A human embryonic limb cell atlas resolved in space and time.
Nature (2023-12-07)
Bao Zhang, Peng He, John E G Lawrence, Shuaiyu Wang, Elizabeth Tuck, Brian A Williams, Kenny Roberts, Vitalii Kleshchevnikov, Lira Mamanova, Liam Bolt, Krzysztof Polanski, Tong Li, Rasa Elmentaite, Eirini S Fasouli, Martin Prete, Xiaoling He, Nadav Yayon, Yixi Fu, Hao Yang, Chen Liang, Hui Zhang, Raphael Blain, Alain Chedotal, David R FitzPatrick, Helen Firth, Andrew Dean, Omer Ali Bayraktar, John C Marioni, Roger A Barker, Mekayla A Storer, Barbara J Wold, Hongbo Zhang, Sarah A Teichmann
ABSTRACT
Human limbs emerge during the fourth post-conception week as mesenchymal buds, which develop into fully formed limbs over the subsequent months1. This process is orchestrated by numerous temporally and spatially restricted gene expression programmes, making congenital alterations in phenotype common2. Decades of work with model organisms have defined the fundamental mechanisms underlying vertebrate limb development, but an in-depth characterization of this process in humans has yet to be performed. Here we detail human embryonic limb development across space and time using single-cell and spatial transcriptomics. We demonstrate extensive diversification of cells from a few multipotent progenitors to myriad differentiated cell states, including several novel cell populations. We uncover two waves of human muscle development, each characterized by different cell states regulated by separate gene expression programmes, and identify musculin (MSC) as a key transcriptional repressor maintaining muscle stem cell identity. Through assembly of multiple anatomically continuous spatial transcriptomic samples using VisiumStitcher, we map cells across a sagittal section of a whole fetal hindlimb. We reveal a clear anatomical segregation between genes linked to brachydactyly and polysyndactyly, and uncover transcriptionally and spatially distinct populations of the mesenchyme in the autopod. Finally, we perform single-cell RNA sequencing on mouse embryonic limbs to facilitate cross-species developmental comparison, finding substantial homology between the two species.
Sigma-Aldrich
Anti-KERA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-PI16 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution