MilliporeSigma
  • Vector-borne Trypanosoma brucei parasites develop in artificial human skin and persist as skin tissue forms.
Vector-borne Trypanosoma brucei parasites develop in artificial human skin and persist as skin tissue forms.
Nature communications (2023-11-24)
Christian Reuter, Laura Hauf, Fabian Imdahl, Rituparno Sen, Ehsan Vafadarnejad, Philipp Fey, Tamara Finger, Nicola G Jones, Heike Walles, Lars Barquist, Antoine-Emmanuel Saliba, Florian Groeber-Becker, Markus Engstler
ABSTRACT
Transmission of Trypanosoma brucei by tsetse flies involves the deposition of the cell cycle-arrested metacyclic life cycle stage into mammalian skin at the site of the fly's bite. We introduce an advanced human skin equivalent and use tsetse flies to naturally infect the skin with trypanosomes. We detail the chronological order of the parasites' development in the skin by single-cell RNA sequencing and find a rapid activation of metacyclic trypanosomes and differentiation to proliferative parasites. Here we show that after the establishment of a proliferative population, the parasites enter a reversible quiescent state characterized by slow replication and a strongly reduced metabolism. We term these quiescent trypanosomes skin tissue forms, a parasite population that may play an important role in maintaining the infection over long time periods and in asymptomatic infected individuals.
Sigma-Aldrich
Anti-KRT14 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution