MilliporeSigma
  • Development and cadherin-mediated control of prefrontal corticostriatal projections in mice.
Development and cadherin-mediated control of prefrontal corticostriatal projections in mice.
iScience (2023-10-19)
Roxana E Mesías, Yosif Zaki, Christopher A Guevara, Lauren G Friedman, Ayan Hussein, Karen Therrien, Alexandra R Magee, Nikolaos Tzavaras, Pamela Del Valle, Mark G Baxter, George W Huntley, Deanna L Benson
ABSTRACT
Action-outcome associations depend on prefrontal cortex (PFC) projections to the dorsal striatum. To assess how these projections form, we measured PFC axon patterning, synapse formation, and functional maturation in the postnatally developing mouse striatum. Using Hotspot analysis, we show that PFC axons form an adult-like pattern of clustered terminations in the first postnatal week that remains largely stable thereafter. PFC-striatal synaptic strength is adult-like by P21, while excitatory synapse density increases until adulthood. We then tested how the targeted deletion of a candidate adhesion/guidance protein, Cadherin-8 (Cdh8), from corticostriatal neurons regulates pathway development. Mutant mice showed diminished PFC axon targeting and reduced spontaneous glutamatergic synaptic activity in the dorsal striatum. They also exhibited impaired behavioral performance in action-outcome learning. The data show that PFC-striatal axons form striatal territories through an early, directed growth model and they highlight essential contributions of Cdh8 to the anatomical and functional features critical for the formation of action-outcome associations.
Product Description
Supelco
Tamoxifen, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Anti-Vesicular Glutamate Transporter 1 Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-β-Catenin Antibody, Upstate®, from rabbit