MilliporeSigma
Carbon dioxide regulates cholesterol levels through SREBP2.
PLoS biology (2023-11-15)
Nityanand Bolshette, Saar Ezagouri, Vaishnavi Dandavate, Iuliia Karavaeva, Marina Golik, Hu Wang, Peter J Espenshade, Timothy F Osborne, Xianlin Han, Gad Asher
ABSTRACT
In mammals, O2 and CO2 levels are tightly regulated and are altered under various pathological conditions. While the molecular mechanisms that participate in O2 sensing are well characterized, little is known regarding the signaling pathways that participate in CO2 signaling and adaptation. Here, we show that CO2 levels control a distinct cellular transcriptional response that differs from mere pH changes. Unexpectedly, we discovered that CO2 regulates the expression of cholesterogenic genes in a SREBP2-dependent manner and modulates cellular cholesterol accumulation. Molecular dissection of the underlying mechanism suggests that CO2 triggers SREBP2 activation through changes in endoplasmic reticulum (ER) membrane cholesterol levels. Collectively, we propose that SREBP2 participates in CO2 signaling and that cellular cholesterol levels can be modulated by CO2 through SREBP2.
Sigma-Aldrich
Anti-SREBP-2 Antibody, clone 22D5, clone 22D5, from rabbit