MilliporeSigma
  • Three-Dimensional Biohybrid StarPEG-Heparin Hydrogel Cultures for Modeling Human Neuronal Development and Alzheimer's Disease Pathology.
Three-Dimensional Biohybrid StarPEG-Heparin Hydrogel Cultures for Modeling Human Neuronal Development and Alzheimer's Disease Pathology.
Methods in molecular biology (Clifton, N.J.) (2022-11-19)
Tohid Siddiqui, Hilal Celikkaya, Zeynep Tansu Atasavum, Stanislava Popova, Uwe Freudenberg, Carsten Werner, Caghan Kizil
ABSTRACT
In this chapter, we present the methodology currently used in our laboratory to generate a starPEG-MMP (starPEG)- and heparin maleimide HM06 (heparin)-based 3D cell culture system, in a hydrogel, that can be used to study human neuronal development and Alzheimer's disease (AD) pathology. A 3D cell culture system can mimic the in vivo cellular environment better than a 2D format, in which these cells exhibit neural network formation, electrophysiological activity, tissue-specific extracellular matrix (ECM) deposition, and neurotransmitter responsiveness. When treated with amyloid beta-42 (Aβ42) peptides, this system recapitulates many of the pathological effects of AD, including reduced neural stem cell proliferation, impaired neuronal network formation, dystrophic axonal ends, synaptic loss, failure to deposit ECM, elevated tau hyperphosphorylation, and formation of neurofibrillary tangles. Culturing human primary cortical astrocyte (pHA)- or induced pluripotent stem cell (iPSC)-derived human neural stem cells in this biohybrid hydrogel system has led to the discovery of novel regulatory pathways underlying neurodegenerative pathology in different phases of AD.
Sigma-Aldrich
Anti-SOX6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse