MilliporeSigma
  • Oleamide, a Sleep-Inducing Supplement, Upregulates Doublecortin in Hippocampal Progenitor Cells via PPARα.
Oleamide, a Sleep-Inducing Supplement, Upregulates Doublecortin in Hippocampal Progenitor Cells via PPARα.
Journal of Alzheimer's disease : JAD (2021-11-09)
Avik Roy, Madhuchhanda Kundu, Sudipta Chakrabarti, Dhruv R Patel, Kalipada Pahan
ABSTRACT
Doublecortin (DCX), a microtubule associated protein, has emerged as a central biomarker of hippocampal neurogenesis. However, molecular mechanisms by which DCX is regulated are poorly understood. Since sleep is involved with the acquisition of memory and oleamide or 9-Octadecenamide (OCT) is a sleep-inducing supplement in human, we examined whether OCT could upregulate DCX in hippocampal progenitor cells (HPCs). We employed real-time PCR, western blot, immunostaining, chromatin immunoprecipitation, lentiviral transduction in HPCs, and the calcium influx assay. OCT directly upregulated the transcription of Dcx in HPCs via activation of peroxisome proliferator-activated receptor α (PPARα), a lipid-lowering transcription factor. We observed that, HPCs of Ppara-null mice displayed significant impairment in DCX expression and neuronal differentiation as compared to that of wild-type mice. Interestingly, treatment with OCT stimulated the differentiation process of HPCs in wild-type, but not Ppara-null mice. Reconstruction of PPARα in mouse Ppara-null HPCs restored the expression of DCX, which was further stimulated with OCT treatment. In contrast, a dominant-negative mutant of PPARα significantly attenuated the stimulatory effect of OCT on DCX expression and suppressed neuronal differentiation of human neural progenitor cells. Furthermore, RNA microarray, STRING, chromatin immunoprecipitation, site-directed mutagenesis, and promoter reporter assay have identified DCX as a new target of PPARα. These results indicate that OCT, a sleep supplement, directly controls the expression of DCX and suggest that OCT may be repurposed for stimulating the hippocampal neurogenesis.
Sigma-Aldrich
Anti-MAP-2, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Microtubule-Associated Protein 2 (MAP2) Antibody, Chemicon®, from rabbit