MilliporeSigma
  • Inhibition of TGF-β signaling, invasion, and growth of cutaneous squamous cell carcinoma by PLX8394.
Inhibition of TGF-β signaling, invasion, and growth of cutaneous squamous cell carcinoma by PLX8394.
Oncogene (2023-10-21)
Elina Siljamäki, Pilvi Riihilä, Ujjwal Suwal, Liisa Nissinen, Pekka Rappu, Markku Kallajoki, Veli-Matti Kähäri, Jyrki Heino
ABSTRACT
Cutaneous squamous cell carcinoma (cSCC) is the most common metastatic skin cancer. The prognosis of patients with metastatic cSCC is poor emphasizing the need for new therapies. We have previously reported that the activation of Ras/MEK/ERK1/2 and transforming growth factor β (TGF-β)/Smad2 signaling in transformed keratinocytes and cSCC cells leads to increased accumulation of laminin-332 and accelerated invasion. Here, we show that the next-generation B-Raf inhibitor PLX8394 blocks TGF-β signaling in ras-transformed metastatic epidermal keratinocytes (RT3 cells) harboring wild-type B-Raf and hyperactive Ras. PLX8394 decreased phosphorylation of TGF-β receptor II and Smad2, as well as p38 activity, MMP-1 and MMP-13 synthesis, and laminin-332 accumulation. PLX8394 significantly inhibited the growth of human cSCC tumors and in vivo collagen degradation in xenograft model. In conclusion, our data indicate that PLX8394 inhibits several serine-threonine kinases in malignantly transformed human keratinocytes and cSCC cells and inhibits cSCC invasion and tumor growth in vitro and in vivo. We identify PLX8394 as a potential therapeutic compound for advanced human cSCC.
Sigma-Aldrich
MMP Inhibitor III, The MMP Inhibitor III, also referenced under CAS 927827-98-3, controls the biological activity of MMP. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.
Sigma-Aldrich
SB 203580, SB 203580, CAS 152121-47-6, is a highly specific, potent, cell-permeable, selective, reversible, and ATP-competitive inhibitor of p38 MAP kinase (IC₅₀ = 34 nM in vitro, 600 nM in cells).
Sigma-Aldrich
PD 98059, PD 98059, CAS 167869-21-8, is a cell-permeable, selective & reversible inhibitor of MAP Kinase Kinase (MEK). Inhibits MAP Kinase activation and subsequent phosphorylation of MAP Kinase substrates.
Sigma-Aldrich
Anti-MMP-13 Antibody, clone 181-15A12, clone 181-15A12, Chemicon®, from mouse
Sigma-Aldrich
LY-294,002 hydrochloride, solid, ≥98% (HPLC)
Sigma-Aldrich
Anti-MMP-1 (Ab-1) Mouse mAb (41-1E5), liquid, clone 41-1E5, Calbiochem®