MilliporeSigma
  • Huntingtin loss in hepatocytes is associated with altered metabolism, adhesion, and liver zonation.
Huntingtin loss in hepatocytes is associated with altered metabolism, adhesion, and liver zonation.
Life science alliance (2023-09-09)
Robert M Bragg, Sydney R Coffey, Jeffrey P Cantle, Shikai Hu, Sucha Singh, Samuel Rw Legg, Cassandra A McHugh, Amreen Toor, Scott O Zeitlin, Seung Kwak, David Howland, Thomas F Vogt, Satdarshan P Monga, Jeffrey B Carroll
ABSTRACT
Huntington's disease arises from a toxic gain of function in the huntingtin (HTT) gene. As a result, many HTT-lowering therapies are being pursued in clinical studies, including those that reduce HTT RNA and protein expression in the liver. To investigate potential impacts, we characterized molecular, cellular, and metabolic impacts of chronic HTT lowering in mouse hepatocytes. Lifelong hepatocyte HTT loss is associated with multiple physiological changes, including increased circulating bile acids, cholesterol and urea, hypoglycemia, and impaired adhesion. HTT loss causes a clear shift in the normal zonal patterns of liver gene expression, such that pericentral gene expression is reduced. These alterations in liver zonation in livers lacking HTT are observed at the transcriptional, histological, and plasma metabolite levels. We have extended these phenotypes physiologically with a metabolic challenge of acetaminophen, for which the HTT loss results in toxicity resistance. Our data reveal an unexpected role for HTT in regulating hepatic zonation, and we find that loss of HTT in hepatocytes mimics the phenotypes caused by impaired hepatic β-catenin function.
Sigma-Aldrich
Anti-CYP2E1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution