MilliporeSigma
  • Inhibition of high level E2F in a RB1 proficient MYCN overexpressing chicken retinoblastoma model normalizes neoplastic behaviour.
Inhibition of high level E2F in a RB1 proficient MYCN overexpressing chicken retinoblastoma model normalizes neoplastic behaviour.
Cellular oncology (Dordrecht) (2023-08-22)
Hanzhao Zhang, Dardan Konjusha, Nima Rafati, Tatsiana Tararuk, Finn Hallböök
ABSTRACT
Retinoblastoma, a childhood cancer, is most frequently caused by bi-allelic inactivation of RB1 gene. However, other oncogenic mutations such as MYCN amplification can induce retinoblastoma with proficient RB1. Previously, we established RB1-proficient MYCN-overexpressing retinoblastoma models both in human organoids and chicken. Here, we investigate the regulatory events in MYCN-induced retinoblastoma carcinogenesis based on the model in chicken. MYCN transformed retinal cells in culture were obtained from in vivo MYCN electroporated chicken embryo retina. The expression profiles were analysed by RNA sequencing. Chemical treatments, qRT-PCR, flow cytometry, immunohisto- and immunocytochemistry and western blot were applied to study the properties and function of these cells. The expression profile of MYCN-transformed retinal cells in culture showed cone photoreceptor progenitor signature and robustly increased levels of E2Fs. This expression profile was consistently observed in long-term culture. Chemical treatments confirmed RB1 proficiency in these cells. The cells were insensitive to p53 activation but inhibition of E2f efficiently induced cell cycle arrest followed by apoptosis. In conclusion, with proficient RB1, MYCN-induced high level of E2F expression dysregulates the cell cycle and contributes to retinoblastoma carcinogenesis. The increased level of E2f renders the cells to adopt a similar mechanistic phenotype to a RB1-deficient tumour.
Sigma-Aldrich
Pifithrin-α, ≥95% (HPLC), powder
Sigma-Aldrich
Nutlin-3a, ≥98% (HPLC)
Sigma-Aldrich
Palbociclib, ≥98% (HPLC)
Sigma-Aldrich
HLM006474, ≥98% (HPLC)