MilliporeSigma
  • Restoring cellular NAD(P)H levels by PPARα and LXRα stimulation to improve mitochondrial complex I deficiency.
Restoring cellular NAD(P)H levels by PPARα and LXRα stimulation to improve mitochondrial complex I deficiency.
Life sciences (2022-04-27)
Sanne J C M Frambach, Ria de Haas, Jan A M Smeitink, Frans G M Russel, Tom J J Schirris
ABSTRACT
Mitochondrial complex I (CI), the first multiprotein enzyme complex of the oxidative phosphorylation system, plays a crucial role in cellular energy production. CI deficiency is associated with a variety of clinical phenotypes, including Leigh syndrome. At the cellular level, an increased NAD(P)H concentration is one of the hallmarks in CI-deficiency. Here, we aimed to attenuate increased NAD(P)H levels by stimulation of ATP-dependent cassette (ABC)A1 and ABCG1-mediated cellular cholesterol efflux with various PPARα and LXRα agonists. Mitochondrial CI-deficient fibroblasts and chemically-induced CI-deficient HeLa cells were used to study the dose-dependent effects of various PPARα and LXRα agonists on cellular NAD(P)H levels and cholesterol efflux. In patient-derived mitochondrial CI-deficient fibroblasts, GW590735, astaxanthin, oleoylethanolamide, and GW3965 significantly reduced the enhanced NAD(P)H levels in CI-deficient fibroblasts. Similar effects were observed in chemically-induced CI-impaired HeLa cells, in which BMS-687453, Wy14643, GW7647, T0901317, DMHCA also demonstrated a beneficial effect. Surprisingly, no effect on ABCA1- and ABCG1-mediated cholesterol efflux in HeLa cells and fibroblasts was found after treatment with these compounds. The reduction in NAD(P)H levels by GW590735 could be partially reversed by inhibition of fatty acid synthase and β-oxidation, which suggests that its beneficial effects are possibly mediated via stimulation of fatty acid metabolism rather than cholesterol efflux. Collectively, PPARα and LXRα stimulation resulted in attenuated cellular NAD(P)H levels in CI-impaired HeLa cells and patient-derived fibroblasts and could eventually have a therapeutic potential in CI deficiency.
Sigma-Aldrich
Apolipoprotein A-I, Human Plasma, High-Density Lipoprotein, Native apolipoprotein A-I from human plasma. Functions as a cofactor for lecithin-cholesterol acyltransferase. A component of high density lipoprotein.
Sigma-Aldrich
Bovine Serum Albumin, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)