MilliporeSigma
  • Role of Epithelial-to-Mesenchymal Transition of Retinal Pigment Epithelial Cells in Glaucoma Cupping.
Role of Epithelial-to-Mesenchymal Transition of Retinal Pigment Epithelial Cells in Glaucoma Cupping.
Journal of clinical medicine (2023-04-14)
Eabha O'Driscoll, Emily Hughes, Mustapha Irnaten, Markus Kuehn, Deborah Wallace, Colm O'Brien
ABSTRACT
Optic nerve head (ONH) cupping is a clinical feature of glaucoma associated with extracellular matrix (ECM) remodelling and lamina cribrosa (LC) fibrosis. Peripapillary atrophy (PPA) occurs commonly in glaucoma, and is characterised by the loss of retinal pigment epithelium (RPE) adjacent to the ONH. Under pro-fibrotic conditions, epithelial cells throughout the body can differentiate into fibroblast-like cells through epithelial-to-mesenchymal transition (EMT) and contribute to ECM fibrosis. This is investigated here in the context of glaucoma and PPA. Human-donor ONH sections were assessed for the presence of the RPE cell-specific marker RPE65 using immunofluorescence. We examined the EMT response of ARPE-19 cells to the following glaucoma-related stimuli: cyclic mechanical stretch, mechanical stiffness, transforming growth factor beta (TGFβ), and tumour necrosis factor alpha (TNFα). The gene expression was measured using the PCR of the epithelial tight junction marker zona occludens 1 (ZO-1) and the mesenchymal markers alpha smooth muscle actin (αSMA) and vimentin. A scratch assay was used to assess the ARPE-19 migration. Significant RPE-65 staining was demonstrated in the glaucomatous ONH. The cyclic stretching and substrate stiffness of the ARPE-19 cells caused a significant decrease in ZO-1 (p = 0.04), and an increase in αSMA (p = 0.04). The scratch assays demonstrated increased migration of ARPE19 in the presence of TNFα (p = 0.02). Furthermore, ARPE-19 cells undergo an EMT-like transition (gain of αSMA, loss of ZO-1 and increased migration) in response to glaucomatous stimuli. This suggests that during PPA, RPE cells have the potential to migrate into the ONH and differentiate into fibroblast-like cells, contributing to glaucomatous ONH cupping.
Sigma-Aldrich
Anti-Mouse IgG (Fab specific) F(ab′)2 fragment–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution