MilliporeSigma
  • A review of biotransformation and pharmacology of ginsenoside compound K.
A review of biotransformation and pharmacology of ginsenoside compound K.
Fitoterapia (2014-12-03)
Xi-Ding Yang, Yong-Yu Yang, Dong-Sheng Ouyang, Guo-Ping Yang
ABSTRACT
As an intestinal bacterial metabolite of ginseng protopanaxadiol saponins, ginsenoside compound K (20-O-beta-d-glucopyranosyl-20(S)-protopanaxadiol, CK) is a major deglycosylated metabolite form of ginsenosides which is absorbed into the systemic circulation. And it has demonstrated such diverse intriguing biological properties as anticarcinogenic, anti-inflammation, antiallergic, anti-diabetic, anti-angiogenesis, anti-aging, neuroprotective and hepatoprotective effects. The present review shall summarize recent studies on various biotransformation and pharmacological activities of CK.
Sigma-Aldrich
Ginsenoside Compound K, ≥96% (HPLC)