MilliporeSigma
  • Idarubicin combats abiraterone and enzalutamide resistance in prostate cells via targeting XPA protein.
Idarubicin combats abiraterone and enzalutamide resistance in prostate cells via targeting XPA protein.
Cell death & disease (2022-12-13)
Ying Zhang, Wei Wei, Changying Li, Siyuan Yan, Shanshan Wang, Shudong Xiao, Chenchen He, Jing Li, Zhi Qi, Benyi Li, Kuo Yang, Changlin Li
ABSTRACT
Although second-generation therapies like abiraterone (ABI) and enzalutamide (ENZ) benefit patients with castration-resistant prostate cancer (CRPC), drug resistance frequently occurs, eventually resulting in therapy failure. In this study, we used two libraries, FDA-approved drug library and CRISP/Cas9 knockout (GeCKO) library to screen for drugs that overcome treatment resistance and to identify the potential drug-resistant genes involved in treatment resistance. Our screening results showed that the DNA-damaging agent idarubicin (IDA) overcame abiraterone and enzalutamide resistance in prostate cancer cells. IDA treatment inhibited the DNA repair protein XPA expression in a transcription-independent manner. Consistently, XPA knockout sensitized prostate cancer cells to abiraterone and enzalutamide treatment. In conclusion, IDA combats abiraterone and enzalutamide resistance by reducing XPA protein level in prostate cancer.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-Androgen Receptor Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-LC3B Antibody, serum, from rabbit