MilliporeSigma
Isolation and Culture of Dorsal Root Ganglia (DRG) from Rodents.
Methods in molecular biology (Clifton, N.J.) (2021-05-26)
Masoud Shekarabi, Jake A Robinson, Tricia H Burdo
ABSTRACT
Preparations of peripheral sensory neurons from rodents are essential for studying the molecular mechanism of neuronal survival and physiology. Although, isolating and culturing these neurons proves difficult, often these preparations are contaminated with nonneuronal proliferating cells. Here, we describe an isolation method using a Percoll gradient and an antimitotic reagent to significantly reduce the nonneuronal cell contamination while maintaining the integrity of the rodent sensory dorsal root ganglia (DRG) neurons.
Sigma-Aldrich
Nerve Growth Factor-2.5S from murine submaxillary gland, NGF-2.5S, lyophilized powder, suitable for cell culture