MilliporeSigma
Evolution and development of parrot pseudoteeth.
Journal of morphology (2021-12-16)
Stéphane Louryan, Myriam Choa-Duterre, Marie Lejong, Nathalie Vanmuylder
ABSTRACT
Parrot embryos carry peculiar appendages at their developing beak that have been described as pseudoteeth. To better characterize the pattern of development responsible for the emergence of these dental appendages, we examined parrot embryos combining conventional histology and microtomography approaches. Using immunohistochemistry, we observed the epithelial and mesenchymal expression of several proteins involved in tooth development in mammals. Parrot pseudoteeth arose by epithelial and mesenchymal evagination, and their early development was similar to the ontogeny of scales and feathers. There was no enamel tissue, and the evaginations were surrounded by the rhamphotheca. In adults, the rhamphotheca covers entirely the appendages, now represented by bone evaginations, which were more numerous in the lower than in the upper beak, being similar to the osseous teeth of the fossil Pelagornithidae. These embryonic pseudoteeth resembled reptile's first-generation teeth and dental appendages of chicken talpid2 mutants. Proteins involved in mammalian odontogenesis, such as SHH, BMP4, PITX2, and PAX9, were found to be generally expressed in beak epithelium and mesenchyme during parrot pseudoteeth development, with clusters of high-level expression in the pseudoteeth rudiments. This suggests that a similar, highly conserved gene expression program gives rise to the appearance of odontode derivatives in numerous species, despite their divergent developmental paths. These results provide new insights into the development and evolution of odontode-derived structures in vertebrates.
Sigma-Aldrich
IHC Select Secondary Goat Anti-Rabbit IgG Antibody, prediluted, biotinylated, from goat, biotin conjugate