MilliporeSigma
  • BioID reveals an ATG9A interaction with ATG13-ATG101 in the degradation of p62/SQSTM1-ubiquitin clusters.
BioID reveals an ATG9A interaction with ATG13-ATG101 in the degradation of p62/SQSTM1-ubiquitin clusters.
EMBO reports (2021-08-10)
Ashari R Kannangara, Daniel M Poole, Colten M McEwan, Joshua C Youngs, Vajira K Weerasekara, Alex M Thornock, Misael T Lazaro, Eranga R Balasooriya, Laura M Oh, Erik J Soderblom, Jonathan J Lee, Daniel L Simmons, Joshua L Andersen
ABSTRACT
ATG9A, the only multi-pass transmembrane protein among core ATG proteins, is an essential regulator of autophagy, yet its regulatory mechanisms and network of interactions are poorly understood. Through quantitative BioID proteomics, we identify a network of ATG9A interactions that includes members of the ULK1 complex and regulators of membrane fusion and vesicle trafficking, including the TRAPP, EARP, GARP, exocyst, AP-1, and AP-4 complexes. These interactions mark pathways of ATG9A trafficking through ER, Golgi, and endosomal systems. In exploring these data, we find that ATG9A interacts with components of the ULK1 complex, particularly ATG13 and ATG101. Using knockout/reconstitution and split-mVenus approaches to capture the ATG13-ATG101 dimer, we find that ATG9A interacts with ATG13-ATG101 independently of ULK1. Deletion of ATG13 or ATG101 causes a shift in ATG9A distribution, resulting in an aberrant accumulation of ATG9A at stalled clusters of p62/SQSTM1 and ubiquitin, which can be rescued by an ULK1 binding-deficient mutant of ATG13. Together, these data reveal ATG9A interactions in vesicle-trafficking and autophagy pathways, including a role for an ULK1-independent ATG13 complex in regulating ATG9A.
Sigma-Aldrich
Monoclonal Anti-PDIA3 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL2444, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Mouse IgG1 (γ1), CF488A antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-EEA1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-TUFM antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL2242, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-VPS26A antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL2287, purified immunoglobulin, buffered aqueous glycerol solution