MilliporeSigma
  • Z-α1-antitrypsin polymers impose molecular filtration in the endoplasmic reticulum after undergoing phase transition to a solid state.
Z-α1-antitrypsin polymers impose molecular filtration in the endoplasmic reticulum after undergoing phase transition to a solid state.
Science advances (2022-04-09)
Joseph E Chambers, Nikita Zubkov, Markéta Kubánková, Jonathon Nixon-Abell, Ioanna Mela, Susana Abreu, Max Schwiening, Giulia Lavarda, Ismael López-Duarte, Jennifer A Dickens, Tomás Torres, Clemens F Kaminski, Liam J Holt, Edward Avezov, James A Huntington, Peter St George-Hyslop, Marina K Kuimova, Stefan J Marciniak
ABSTRACT
Misfolding of secretory proteins in the endoplasmic reticulum (ER) features in many human diseases. In α1-antitrypsin deficiency, the pathogenic Z variant aberrantly assembles into polymers in the hepatocyte ER, leading to cirrhosis. We show that α1-antitrypsin polymers undergo a liquid:solid phase transition, forming a protein matrix that retards mobility of ER proteins by size-dependent molecular filtration. The Z-α1-antitrypsin phase transition is promoted during ER stress by an ATF6-mediated unfolded protein response. Furthermore, the ER chaperone calreticulin promotes Z-α1-antitrypsin solidification and increases protein matrix stiffness. Single-particle tracking reveals that solidification initiates in cells with normal ER morphology, previously assumed to represent a healthy pool. We show that Z-α1-antitrypsin-induced hypersensitivity to ER stress can be explained by immobilization of ER chaperones within the polymer matrix. This previously unidentified mechanism of ER dysfunction provides a template for understanding a diverse group of related proteinopathies and identifies ER chaperones as potential therapeutic targets.
Sigma-Aldrich
Anti-α1-Antitrypsin antibody produced in rabbit, IgG fraction of antiserum