MilliporeSigma
  • Valosin-containing protein immunoreactivity in tauopathies, synucleinopathies, polyglutamine diseases and intranuclear inclusion body disease.
Valosin-containing protein immunoreactivity in tauopathies, synucleinopathies, polyglutamine diseases and intranuclear inclusion body disease.
Neuropathology : official journal of the Japanese Society of Neuropathology (2013-06-21)
Fumiaki Mori, Kunikazu Tanji, Yasuko Toyoshima, Hidenao Sasaki, Mari Yoshida, Akiyoshi Kakita, Hitoshi Takahashi, Koichi Wakabayashi
ABSTRACT
Valosin-containing protein (VCP) is associated with multiple cellular functions, including ubiquitin-dependent protein degradation. Mutations in VCP are known to cause inclusion body myopathy with Paget's disease and frontotemporal dementia and familial amyotrophic lateral sclerosis (fALS; ALS14), both of which are characterized by trans-activation response DNA protein 43 (TDP-43)-positive neuronal cytoplasmic and nuclear inclusions. Recently, immunoreactivity for fALS-associated proteins (TDP-43, fused in sarcoma (FUS), optineurin and ubiquilin-2) were reported to be present in cytoplasmic and nuclear inclusions in various neurodegenerative diseases. However, the extent and frequency of VCP-immunoreactive structures in these neurodegenerative diseases are uncertain. We immunohistochemically examined the brains of 72 cases with neurodegenerative diseases and five control cases. VCP immunoreactivity was present in Lewy bodies in Parkinson's disease and dementia with Lewy bodies, and neuronal nuclear inclusions in five polyglutamine diseases and intranuclear inclusion body disease, as well as in Marinesco bodies in aged control subjects. However, other neuronal and glial cytoplasmic inclusions in tauopathies and TDP-43 proteinopathies were unstained. These findings suggest that VCP may have common mechanisms in the formation or degradation of cytoplasmic and nuclear inclusions of neurons, but not of glial cells, in several neurodegenerative conditions.
Sigma-Aldrich
Anti-VCP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution