MilliporeSigma
  • Comprehensive, Integrative Genomic Analysis of Diffuse Lower-Grade Gliomas.
Comprehensive, Integrative Genomic Analysis of Diffuse Lower-Grade Gliomas.
The New England journal of medicine (2015-06-11)
Daniel J Brat, Roel G W Verhaak, Kenneth D Aldape, W K Alfred Yung, Sofie R Salama, Lee A D Cooper, Esther Rheinbay, C Ryan Miller, Mark Vitucci, Olena Morozova, A Gordon Robertson, Houtan Noushmehr, Peter W Laird, Andrew D Cherniack, Rehan Akbani, Jason T Huse, Giovanni Ciriello, Laila M Poisson, Jill S Barnholtz-Sloan, Mitchel S Berger, Cameron Brennan, Rivka R Colen, Howard Colman, Adam E Flanders, Caterina Giannini, Mia Grifford, Antonio Iavarone, Rajan Jain, Isaac Joseph, Jaegil Kim, Katayoon Kasaian, Tom Mikkelsen, Bradley A Murray, Brian Patrick O'Neill, Lior Pachter, Donald W Parsons, Carrie Sougnez, Erik P Sulman, Scott R Vandenberg, Erwin G Van Meir, Andreas von Deimling, Hailei Zhang, Daniel Crain, Kevin Lau, David Mallery, Scott Morris, Joseph Paulauskis, Robert Penny, Troy Shelton, Mark Sherman, Peggy Yena, Aaron Black, Jay Bowen, Katie Dicostanzo, Julie Gastier-Foster, Kristen M Leraas, Tara M Lichtenberg, Christopher R Pierson, Nilsa C Ramirez, Cynthia Taylor, Stephanie Weaver, Lisa Wise, Erik Zmuda, Tanja Davidsen, John A Demchok, Greg Eley, Martin L Ferguson, Carolyn M Hutter, Kenna R Mills Shaw, Bradley A Ozenberger, Margi Sheth, Heidi J Sofia, Roy Tarnuzzer, Zhining Wang, Liming Yang, Jean Claude Zenklusen, Brenda Ayala, Julien Baboud, Sudha Chudamani, Mark A Jensen, Jia Liu, Todd Pihl, Rohini Raman, Yunhu Wan, Ye Wu, Adrian Ally, J Todd Auman, Miruna Balasundaram, Saianand Balu, Stephen B Baylin, Rameen Beroukhim, Moiz S Bootwalla, Reanne Bowlby, Christopher A Bristow, Denise Brooks, Yaron Butterfield, Rebecca Carlsen, Scott Carter, Lynda Chin, Andy Chu
ABSTRACT
Diffuse low-grade and intermediate-grade gliomas (which together make up the lower-grade gliomas, World Health Organization grades II and III) have highly variable clinical behavior that is not adequately predicted on the basis of histologic class. Some are indolent; others quickly progress to glioblastoma. The uncertainty is compounded by interobserver variability in histologic diagnosis. Mutations in IDH, TP53, and ATRX and codeletion of chromosome arms 1p and 19q (1p/19q codeletion) have been implicated as clinically relevant markers of lower-grade gliomas. We performed genomewide analyses of 293 lower-grade gliomas from adults, incorporating exome sequence, DNA copy number, DNA methylation, messenger RNA expression, microRNA expression, and targeted protein expression. These data were integrated and tested for correlation with clinical outcomes. Unsupervised clustering of mutations and data from RNA, DNA-copy-number, and DNA-methylation platforms uncovered concordant classification of three robust, nonoverlapping, prognostically significant subtypes of lower-grade glioma that were captured more accurately by IDH, 1p/19q, and TP53 status than by histologic class. Patients who had lower-grade gliomas with an IDH mutation and 1p/19q codeletion had the most favorable clinical outcomes. Their gliomas harbored mutations in CIC, FUBP1, NOTCH1, and the TERT promoter. Nearly all lower-grade gliomas with IDH mutations and no 1p/19q codeletion had mutations in TP53 (94%) and ATRX inactivation (86%). The large majority of lower-grade gliomas without an IDH mutation had genomic aberrations and clinical behavior strikingly similar to those found in primary glioblastoma. The integration of genomewide data from multiple platforms delineated three molecular classes of lower-grade gliomas that were more concordant with IDH, 1p/19q, and TP53 status than with histologic class. Lower-grade gliomas with an IDH mutation either had 1p/19q codeletion or carried a TP53 mutation. Most lower-grade gliomas without an IDH mutation were molecularly and clinically similar to glioblastoma. (Funded by the National Institutes of Health.).
Sigma-Aldrich
ATRX Rabbit Polyclonal Antibody