MilliporeSigma
  • Differential role of tumor necrosis factor receptors in mouse brain inflammatory responses in cryolesion brain injury.
Differential role of tumor necrosis factor receptors in mouse brain inflammatory responses in cryolesion brain injury.
Journal of neuroscience research (2005-11-04)
Albert Quintana, Mercedes Giralt, Santiago Rojas, Milena Penkowa, Iain L Campbell, Juan Hidalgo, Amalia Molinero
ABSTRACT
Tumor necrosis factor-alpha (TNF-alpha) is one of the mediators dramatically increased after traumatic brain injury that leads to the activation, proliferation, and hypertrophy of mononuclear, phagocytic cells and gliosis. Eventually, TNF-alpha can induce both apoptosis and necrosis via intracellular signaling. This cytokine exerts its functions via interaction with two receptors: type-1 receptor (TNFR1) and type-2 receptor (TNFR2). In this work, the inflammatory response after a freeze injury (cryolesion) in the cortex was studied in wild-type (WT) animals and in mice lacking TNFR1 (TNFR1 KO) or TNFR2 (TNFR2 KO). Lack of TNFR1, but not of TNFR2, significantly decreased the inflammatory response and tissue damage elicited by the cryolesion at both 3 and 7 days postlesion, with decreased gliosis, lower IL-1beta immunostaining, and a reduction of apoptosis markers. Cryolesion produced a clear induction of the proinflammatory cytokines interleukin (IL)-1alpha, IL-1beta, IL-6, and TNF-alpha; this induction was significantly lower in the TNFR1 KO mice. Host response genes (ICAM-1, A20, EB22/5, and GFAP) were also induced by the cryolesion, but to a lesser extent in TNFR1 KO mice. Lack of TNFR1 signaling also affected the expression of apoptosis/cell death-related genes (Fas, Rip, p53), matrix metalloproteinases (MMP3, MMP9, MMP12), and their inhibitors (TIMP1), suggesting a role of TNFR1 in extracellular matrix remodeling after injury. However, GDNF, NGF, and BDNF expression were not affected by TNFR1 deficiency. Overall, these results suggest that TNFR1 is involved in the early establishment of the inflammatory response and that its deficiency causes a decreased inflammatory response and tissue damage following brain injury.
Sigma-Aldrich
Monoclonal Anti-Rabbit Immunoglobulins–Biotin antibody produced in mouse, clone RG-16, purified immunoglobulin, buffered aqueous solution