MilliporeSigma
  • 5-HT3A receptors maintain hippocampal LTP in a CB1 and GABAA receptor- dependent manner for spatial memory.
5-HT3A receptors maintain hippocampal LTP in a CB1 and GABAA receptor- dependent manner for spatial memory.
British journal of pharmacology (2022-01-09)
Yan Yu, Jing-Jing Li, Xiao-Qian He, Zi-Ying Lai, Rui Hao, Yu Qi, Dong-Qing Cao, Ming Fu, Hong Ma, Qiu-Chen Xie, Mu Sun, Zhi-Li Huang, Ling-Jing Jin, Hui-Hui Sun, Ning Lu, Rui Wang, Wing-Ho Yung, Ying Huang
ABSTRACT
As the only ionotropic receptor in the 5-HT receptor family, the 5-HT3 receptor (5-HT3 R) is involved in psychiatric disorders and its modulators have potential therapeutic effects for cognitive impairment in these disorders. However, it remains unclear how 5-HT3 Rs shape synaptic plasticity for memory function. Extracellular as well as whole-cell electrophysiological recordings were used to monitor hippocampal LTP and synaptic transmission in hippocampal slices in 5-HT3 AR knockout or 5-HT3 AR-GFP mice. Immunocytochemistry, qRT-PCR and western blotting were used to measure receptor expression. We also assessed hippocampal dependent cognition and memory, using the Morris water maze (MWM) and novel object recognition. We found that 5-HT3 R dysfunction impaired hippocampal LTP in Schaffer collateral (SC)-CA1 pathway in hippocampal slices, by facilitating GABAergic inputs in pyramidal cells. This effect was dependent on 5-HT3 Rs on axon terminals. It resulted from reduced expression and function of the cannabinoid receptor 1 (CB1 R) co-localized with 5-HT3 Rs on axon terminals, and then led to diminishment of tonic inhibition of GABA release by CB1 Rs. Inhibition of CB1 Rs mimicked the facilitation of GABAergic transmission by 5-HT3 R disruption. Consequently, mice with hippocampal 5-HT3 R disruption exhibited impaired spatial memory in MWM tasks. These results suggest that 5-HT3 Rs are crucial in enabling hippocampal synaptic plasticity via a novel CB1 R-GABAA -dependent pathway to regulate spatial memory.
Sigma-Aldrich
Anti-CNR1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Cholecystokinin (26-33) (CCK-8) antibody produced in rabbit, whole antiserum