MilliporeSigma
  • Circ_0075960 targets the miR-202-5p/CTNND1 axis to promote the growth and migration of endometrial carcinoma cells via regulating Wnt/β-catenin signaling activity.
Circ_0075960 targets the miR-202-5p/CTNND1 axis to promote the growth and migration of endometrial carcinoma cells via regulating Wnt/β-catenin signaling activity.
Journal of gynecologic oncology (2022-11-26)
Yan Nian, Xiaorong Li, Jingwen Ma, Ting Gao, Dan Liu
ABSTRACT
Endometrial carcinoma (EC) is one of the most common malignant tumors of the female reproductive tract, involving multiple molecular alterations. Circular RNA (circRNA) dysregulation is frequently observed in EC tissues, suggesting the involvement of circRNA in EC development. We aimed to investigate the role of circ_0075960 in EC. Real-time quantitative polymerase chain reaction (RT-qPCR) and western blot assays were applied for expression analysis. CCK-8, EdU, colony formation, flow cytometry and wound healing assays were employed for functional analysis. The predicted binding relationship between miR-202-5p and circ_0075960 or CTNND1 was validated by dual-luciferase reporter experiment. In vivo animal models were constructed in nude mice to verify the role of circ_0075960 in tumor growth. Circ_0075960 and CTNND1 were upregulated, while miR-202-5p was downregulated in EC. Knockdown of circ_0075960 induced EC cell apoptosis, suppressed cell proliferation and migration, and repressed tumor growth in animal models. MiR-202-5p was targeted by circ_0075960 and it directly bound to CTNND1 3'UTR. The inhibition of circ_0075960 knockdown or miR-202-5p enrichment on EC cell proliferation and migration was reversed by miR-202-5p depletion or CTNND1 overexpression, respectively. Circ_0075960 targeted miR-202-5p to positively regulate CTNND1 expression. Moreover, circ_0075960 knockdown weakened the activity of Wnt/β-catenin signaling via targeting the miR-202-5p/CTNND1 axis. Circ_0075960 targets the miR-202-5p/CTNND1 axis to modulate Wnt/β-catenin signaling activity, thus contributing to the malignant development of EC.
Sigma-Aldrich
Anti-CTNND1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-CTNND1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution