MilliporeSigma
  • Activation of RhoC by regulatory ubiquitination is mediated by LNX1 and suppressed by LIS1.
Activation of RhoC by regulatory ubiquitination is mediated by LNX1 and suppressed by LIS1.
Scientific reports (2022-10-04)
Stanislav Kholmanskikh, Shawn Singh, M Elizabeth Ross
ABSTRACT
Regulation of Rho GTPases remains a topic of active investigation as they are essential participants in cell biology and the pathophysiology of many human diseases. Non-degrading ubiquitination (NDU) is a critical regulator of the Ras superfamily, but its relevance to Rho proteins remains unknown. We show that RhoC, but not RhoA, is a target of NDU by E3 ubiquitin ligase, LNX1. Furthermore, LNX1 ubiquitination of RhoC is negatively regulated by LIS1 (aka, PAFAH1B1). Despite multiple reports of functional interaction between LIS1 and activity of Rho proteins, a robust mechanism linking the two has been lacking. Here, LIS1 inhibition of LNX1 effects on RhoGDI-RhoC interaction provides a molecular mechanism underpinning the enhanced activity of Rho proteins observed upon reduction in LIS1 protein levels. Since LNX1 and RhoC are only found in vertebrates, the LIS1-LNX1-RhoC module represents an evolutionarily acquired function of the highly conserved LIS1. While these nearly identical proteins have several distinct RhoA and RhoC downstream effectors, our data provide a rare example of Rho-isoform specific, upstream regulation that opens new therapeutic opportunities.
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Anti-LIS1 antibody, Mouse monoclonal, clone LIS1-338, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Rho (-A, -B, -C) Antibody, clone 3L74, rabbit monoclonal, clone 3L74, Upstate®, from rabbit
Sigma-Aldrich
Gelatin from cold water fish skin, solid
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Millipore
EZview Red ANTI-FLAG® M2 Affinity Gel, clone M2