MilliporeSigma
  • Antibodies to biotin enable large-scale detection of biotinylation sites on proteins.
Antibodies to biotin enable large-scale detection of biotinylation sites on proteins.
Nature methods (2017-10-19)
Namrata D Udeshi, Kayvon Pedram, Tanya Svinkina, Shaunt Fereshetian, Samuel A Myers, Ozan Aygun, Karsten Krug, Karl Clauser, Dominic Ryan, Tslil Ast, Vamsi K Mootha, Alice Y Ting, Steven A Carr
ABSTRACT
Although purification of biotinylated molecules is highly efficient, identifying specific sites of biotinylation remains challenging. We show that anti-biotin antibodies enable unprecedented enrichment of biotinylated peptides from complex peptide mixtures. Live-cell proximity labeling using APEX peroxidase followed by anti-biotin enrichment and mass spectrometry yielded over 1,600 biotinylation sites on hundreds of proteins, an increase of more than 30-fold in the number of biotinylation sites identified compared to streptavidin-based enrichment of proteins.
Sigma-Aldrich
Monoclonal Anti-Biotin–Agarose antibody produced in mouse, clone BN-34, purified immunoglobulin, PBS suspension