MilliporeSigma
  • NO-Dependent Mechanisms of p53 Expression and Cell Death in Rat's Dorsal Root Ganglia after Sciatic-Nerve Transection.
NO-Dependent Mechanisms of p53 Expression and Cell Death in Rat's Dorsal Root Ganglia after Sciatic-Nerve Transection.
Biomedicines (2022-07-28)
Stanislav Rodkin, Valentina Dzreyan, Mikhail Bibov, Alexey Ermakov, Tatyana Derezina, Evgeniya Kirichenko
ABSTRACT
Peripheral-nerve injury is a frequent cause of disability. Presently, no clinically effective neuroprotectors have been found. We have studied the NO-dependent expression of p53 in the neurons and glial cells of the dorsal root ganglia (DRG) of a rat's spinal cord, as well as the role of NO in the death of these cells under the conditions of axonal stress, using sciatic-nerve axotomy as a model. It was found out that axotomy led to the nuclear-cytoplasmic redistribution of p53 in neurons, 24 h after trauma. The NO donor led to a considerable increase in the level of p53 in nuclei and, to a smaller degree, in the cytoplasm of neurons and karyoplasm of glial cells 4 and 24 h after axotomy. Application of a selective inhibitor of inducible NO-synthase (iNOS) provided the opposite effect. Introduction of the NO donor resulted in a significant increase in cell death in the injured ipsilateral DRG, 24 h and 7 days after trauma. The selective inhibitor of iNOS demonstrated a neuroprotective effect. Axotomy was shown to upregulate the iNOS in nuclei and cytoplasm of DRG cells. The NO-dependent expression of p53, which is particularly achieved through iNOS activation, is believed to be a putative signaling mechanism of neural and glial-cell death after axotomy.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-Mouse IgG1 (γ1), CF555 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Nitric Oxide Synthase, Inducible antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
NuCLEAR Extraction Kit, For mammalian tissue or cultured cells
Millipore
Benzonase® Nuclease, ≥250 units/μL, ≥90% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-p53 antibody, Mouse monoclonal, clone BP53-12, ascites fluid
Sigma-Aldrich
Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF 488A antibody produced in goat, ~2 mg/mL, affinity isolated antibody
Sigma-Aldrich
Anti-NeuN Antibody (rabbit), from rabbit, purified by affinity chromatography