MilliporeSigma
  • ITGβ6 Facilitates Skeletal Muscle Development by Maintaining the Properties and Cytoskeleton Stability of Satellite Cells.
ITGβ6 Facilitates Skeletal Muscle Development by Maintaining the Properties and Cytoskeleton Stability of Satellite Cells.
Life (Basel, Switzerland) (2022-07-28)
Hong Zhang, Yuan Liu, Cencen Li, Weiya Zhang
ABSTRACT
Integrin proteins are important receptors connecting the intracellular skeleton of satellite cells and the extracellular matrix (ECM), playing an important role in the process of skeletal muscle development. In this research, the function of ITGβ6 in regulating the differentiation of satellite cells was studied. Transcriptome and proteome analysis indicated that Itgβ6 is a key node connecting ECM-related proteins to the cytoskeleton, and it is necessary for the integrity of the membrane structure and stability of the cytoskeletal system, which are essential for satellite cell adhesion. Functional analysis revealed that the ITGβ6 protein could affect the myogenic differentiation potential of satellite cells by regulating the expression of PAX7 protein, thus regulating the formation of myotubes. Moreover, ITGβ6 is involved in muscle development by regulating cell-adhesion-related proteins, such as β-laminin, and cytoskeletal proteins such as PXN, DMD, and VCL. In conclusion, the effect of ITGβ6 on satellite cell differentiation mainly occurs before the initiation of differentiation, and it regulates terminal differentiation by affecting satellite cell characteristics, cell adhesion, and the stability of the cytoskeleton system.
Sigma-Aldrich
Collagenase from Clostridium histolyticum, 0.2 μm filtered, for general use, Type I-S, 0.2-1.0 FALGPA units/mg solid, ≥125 CDU/mg solid