MilliporeSigma
  • Rapid Detection and Inhibition of SARS-CoV-2-Spike Mutation-Mediated Microthrombosis.
Rapid Detection and Inhibition of SARS-CoV-2-Spike Mutation-Mediated Microthrombosis.
Advanced science (Weinheim, Baden-Wurttemberg, Germany) (2021-10-24)
Sandro Satta, Angela Lai, Susana Cavallero, Cayden Williamson, Justin Chen, Ana M Blázquez-Medela, Mehrdad Roustaei, Barbara J Dillon, Nureddin Ashammakhi, Dino Di Carlo, Zhaoping Li, Ren Sun, Tzung K Hsiai
ABSTRACT
Activation of endothelial cells following severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) infection is thought to be the primary driver for the increasingly recognized thrombotic complications in coronavirus disease 2019 patients, potentially due to the SARS-CoV-2 Spike protein binding to the human angiotensin-converting enzyme 2 (hACE2). Vaccination therapies use the same Spike sequence or protein to boost host immune response as a protective mechanism against SARS-CoV-2 infection. As a result, cases of thrombotic events are reported following vaccination. Although vaccines are generally considered safe, due to genetic heterogeneity, age, or the presence of comorbidities in the population worldwide, the prediction of severe adverse outcome in patients remains a challenge. To elucidate Spike proteins underlying patient-specific-vascular thrombosis, the human microcirculation environment is recapitulated using a novel microfluidic platform coated with human endothelial cells and exposed to patient specific whole blood. Here, the blood coagulation effect is tested after exposure to Spike protein in nanoparticles and Spike variant D614G in viral vectors and the results are corroborated using live SARS-CoV-2. Of note, two potential strategies are also examined to reduce blood clot formation, by using nanoliposome-hACE2 and anti-Interleukin (IL) 6 antibodies.
Sigma-Aldrich
Anti-Mouse IgG1 (γ1), CF633 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Fibrinogen antibody produced in mouse, clone 85D4, ascites fluid