MilliporeSigma
Imine as a linchpin approach for meta-C-H functionalization.
Nature communications (2021-03-04)
Sukdev Bag, Sadhan Jana, Sukumar Pradhan, Suman Bhowmick, Nupur Goswami, Soumya Kumar Sinha, Debabrata Maiti
ABSTRACT
Despite the widespread applications of C-H functionalization, controlling site selectivity remains a significant challenge. Covalently attached directing groups (DGs) served as ancillary ligands to ensure ortho-, meta- and para-C-H functionalization over the last two decades. These covalently linked DGs necessitate two extra steps for a single C-H functionalization: introduction of DG prior to C-H activation and removal of DG post-functionalization. Here we report a temporary directing group (TDG) for meta-C-H functionalization via reversible imine formation. By overruling facile ortho-C-H bond activation by imine-N atom, a suitably designed pyrimidine-based TDG successfully delivered selective meta-C-C bond formation. Application of this temporary directing group strategy for streamlining the synthesis of complex organic molecules without any necessary pre-functionalization at the meta position has been explored.
Sigma-Aldrich
2-fluoro-6-(pyrimidin-5-yl)aniline, ≥95%
Sigma-Aldrich
2-(Pyrimidin-5-yl)benzaldehyde