MilliporeSigma
  • Serum bridging molecules drive candidal invasion of human but not mouse endothelial cells.
Serum bridging molecules drive candidal invasion of human but not mouse endothelial cells.
PLoS pathogens (2022-07-08)
Quynh T Phan, Norma V Solis, Jianfeng Lin, Marc Swidergall, Shakti Singh, Hong Liu, Donald C Sheppard, Ashraf S Ibrahim, Aaron P Mitchell, Scott G Filler
ABSTRACT
During hematogenously disseminated candidiasis, blood borne fungi must invade the endothelial cells that line the blood vessels to infect the deep tissues. Although Candida albicans, which forms hyphae, readily invades endothelial cells, other medically important species of Candida are poorly invasive in standard in vitro assays and have low virulence in immunocompetent mouse models of disseminated infection. Here, we show that Candida glabrata, Candida tropicalis, Candida parapsilosis, and Candida krusei can bind to vitronectin and high molecular weight kininogen present in human serum. Acting as bridging molecules, vitronectin and kininogen bind to αv integrins and the globular C1q receptor (gC1qR), inducing human endothelial cells to endocytose the fungus. This mechanism of endothelial cell invasion is poorly supported by mouse endothelial cells but can be restored when mouse endothelial cells are engineered to express human gC1qR or αv integrin. Overall, these data indicate that bridging molecule-mediated endocytosis is a common pathogenic strategy used by many medically important Candida spp. to invade human vascular endothelial cells.
Sigma-Aldrich
Anti-Integrin αVβ5 Antibody, clone P1F6, clone P1F6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin α5 Antibody, CT, Intracellular, serum, Chemicon®
Sigma-Aldrich
Anti-Integrin αVβ3 Antibody, clone LM609, clone LM609, Chemicon®, from mouse
Sigma-Aldrich
Anti-Vitronectin Antibody, clone 8E6(LJ8), clone 8E6(LJ8), Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin alphaV Antibody, CT, Intracellular, serum, Chemicon®