MilliporeSigma
EGCG Restricts PRRSV Proliferation by Disturbing Lipid Metabolism.
Microbiology spectrum (2022-04-12)
Peng-Wei Yu, Peng-Fei Fu, Lei Zeng, Yan-Li Qi, Xiu-Qing Li, Qi Wang, Guo-Yu Yang, Hua-Wei Li, Jiang Wang, Bei-Bei Chu, Meng-Di Wang
ABSTRACT
Porcine reproductive and respiratory syndrome virus (PRRSV) infection leads to late-term reproductive failure and respiratory illness that affect the global swine industry. Epigallocatechin gallate (EGCG) is a polyphenolic compound from green tea that exerts antiviral activity against diverse viruses. This study aimed to report an uncharacterized mechanism of how EGCG restricted PRRSV proliferation. EGCG showed no significant effects on cell viability, cell cycle progression, and apoptosis in porcine alveolar macrophages and MARC-145 cells. The treatment of cells with EGCG attenuated the replication of both highly pathogenic and less pathogenic PRRSV in vitro. The viral life cycle analysis demonstrated that EGCG affected PRRSV replication and assembly, but not viral attachment, entry, or release. Interestingly, EGCG treatment abrogated the increased lipid droplets formation and lipid content induced by PRRSV infection. We further demonstrated that EGCG blocked PRRSV-stimulated expression of the key enzymes in lipid synthesis. In addition, EGCG attenuated PRRSV-induced autophagy that is critical for PRRSV proliferation. The supplementation of oleic acid restored PRRSV replication and assembly under EGCG treatment. Together, our results support that EGCG inhibits PRRSV proliferation through disturbing lipid metabolism. IMPORTANCE Porcine reproductive and respiratory syndrome virus (PRRSV) is an enveloped single-positive-stranded RNA virus that causes acute respiratory distress in piglets and reproductive failure in sows, resulting in huge economic losses to the global swine industry. Several lines of evidence have suggested the crucial roles of lipids in PRRSV proliferation. Our previous report demonstrated that PRRSV activated lipophagy to facilitate viral replication through downregulating the expression of N-Myc downstream-regulated gene 1. The manipulation of lipid metabolism may be a new perspective to prevent PRRSV spread. In the present study, we reported that epigallocatechin-3-gallate (EGCG), the major component of green tea catechins, significantly attenuated PRRSV infection through inhibiting lipid synthesis and autophagy. Given that natural products derived from plants have helped in the prevention and treatment of various infectious diseases, EGCG has a great potential to serve as a safe and environmentally friendly natural compound to treat PRRSV infection.
Sigma-Aldrich
Anti-HMG-CoA Reductase Antibody, clone IgG-A9, clone IgG-A9, from mouse
Sigma-Aldrich
Monoclonal Anti-Cy3/Cy5−Biotin antibody produced in mouse, ~1 mg/mL, clone CY-96, purified immunoglobulin, buffered aqueous solution