MilliporeSigma
  • AMP activated kinase negatively regulates hepatic Fetuin-A via p38 MAPK-C/EBPβ/E3 Ubiquitin Ligase Signaling pathway.
AMP activated kinase negatively regulates hepatic Fetuin-A via p38 MAPK-C/EBPβ/E3 Ubiquitin Ligase Signaling pathway.
PloS one (2022-05-07)
Vishal Kothari, Jeganathan Ramesh Babu, Suresh T Mathews
ABSTRACT
Fetuin-A (Fet-A) is a liver-secreted phosphorylated protein, known to impair insulin signaling, which has been shown to be associated with obesity, insulin resistance, and incident diabetes. Fet-A interacts with the insulin-stimulated insulin receptor (IR) and inhibits IR tyrosine kinase activity and glucose uptake. It has been shown that high glucose increases Fet-A expression through the ERK1/2 signaling pathway. However, factors that downregulate Fet-A expression and their potential mechanisms are unclear. We examined the effect of AMP-activated protein kinase (AMPK) on high-glucose induced Fet-A expression in HepG2 cells, Hep3B cells and primary rat hepatocytes. High glucose increased Fet-A and phosphorylated (Ser312) fetuin-A (pFet-A) expression, which are known to impair insulin signaling. AICAR-induced AMPK activation significantly down-regulated high glucose-induced Fet-A expression and secretion of pFet-A while treatment with Compound C (AMPK inhibitor), SB202190 (p38 MAPK inhibitor) or p38 MAPK siRNA transfection prevented AICAR-induced downregulation of Fet-A expression. In addition, activation of p38 MAPK, by anisomycin, decreased the hepatic expression of Fet-A. Further, we our studies have shown that short-term effect of AICAR-treatment on Fet-A expression was mediated by proteosomal degradation, and long-term treatment of AICAR was associated with decrease in hepatic expression of C/EBP beta, an important transcription factor involved in the regulation of Fet-A. Taken together, our studies implicate a critical role for AMPK-p38 MAPK-C/EBPb-ubiquitin-proteosomal axis in the regulation of the expression of hepatic Fet-A.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Fetal Bovine Serum, USA origin, suitable for cell culture
Protein A-Sepharose CL-4B, Cytiva 17-0780-01, pack of 1.5 g