MilliporeSigma
  • Vascular smooth muscle cell growth arrest on blockade of thrombospondin-1 requires p21(Cip1/WAF1).
Vascular smooth muscle cell growth arrest on blockade of thrombospondin-1 requires p21(Cip1/WAF1).
The American journal of physiology (1999-09-14)
D Chen, K Guo, J Yang, W A Frazier, J M Isner, V Andrés
ABSTRACT
Abnormal proliferation of vascular smooth muscle cells (VSMCs) is thought to play an important role in the pathogenesis of atherosclerosis and restenosis. Previous studies have implicated the extracellular matrix protein thrombospondin-1 (TSP1) in mitogen-dependent proliferation of VSMCs. In this study, we investigated the molecular mechanisms involved in TSP1-mediated regulation of VSMC growth. Neutralizing A4.1 anti-TSP1 antibody inhibited the activity of the G(1)/S cyclin-dependent kinase 2 (cdk2) and blocked the induction of S-phase entry, which normally occurs in serum-stimulated VSMCs. This growth-inhibitory effect was associated with a marked induction of p21(Cip1/WAF1) (p21) expression in A4.1-treated VSMCs. Moreover, addition of A4.1 antibody to VSMCs markedly increased the level of p21 bound to cdk2. Thus growth arrest on antibody blockade of TSP1 may be mediated by the cdk inhibitory protein p21. Consistent with this notion, anti-TSP1 antibody inhibited [(3)H]-thymidine incorporation in wild-type but not in p21-deficient mouse embryonic fibroblasts (MEFs). Together, these data suggest that p21 plays an important role in TSP1-mediated control of cellular proliferation.
Sigma-Aldrich
IgM, Lambda from murine myeloma, clone MOPC 104E, ascites fluid, lyophilized powder