MilliporeSigma
  • Protocol for Purification of Human ZGRF1 and Its Regulatory Function on RAD51-Mediated D-Loop Formation.
Protocol for Purification of Human ZGRF1 and Its Regulatory Function on RAD51-Mediated D-Loop Formation.
STAR protocols (2020-09-29)
Michael Lisby, Xiaoyu Xue
ABSTRACT
Human ZGRF1 is one of the many helicases that play important roles in the repair of damaged chromosomes by homologous recombination. However, the lack of biochemical characterization of the ZGRF1 protein has hindered the understanding of its function. We have developed a facile protocol to express human ZGRF1 in insect cells and for its purification. We have also tuned biochemical assays by lowering the protein and DNA concentrations to analyze its functions in RAD51-mediated D-loop formation and dissociation. For complete details on the use and execution of this protocol, please refer to Brannvoll et al. (2020).
Sigma-Aldrich
Rosetta(DE3) Competent Cells - Novagen, Rosetta host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli.