MilliporeSigma
  • SHLD1 is dispensable for 53BP1-dependent V(D)J recombination but critical for productive class switch recombination.
SHLD1 is dispensable for 53BP1-dependent V(D)J recombination but critical for productive class switch recombination.
Nature communications (2022-06-29)
Estelle Vincendeau, Wenming Wei, Xuefei Zhang, Cyril Planchais, Wei Yu, Hélène Lenden-Hasse, Thomas Cokelaer, Juliana Pipoli da Fonseca, Hugo Mouquet, David J Adams, Frederick W Alt, Stephen P Jackson, Gabriel Balmus, Chloé Lescale, Ludovic Deriano
ABSTRACT
SHLD1 is part of the Shieldin (SHLD) complex, which acts downstream of 53BP1 to counteract DNA double-strand break (DSB) end resection and promote DNA repair via non-homologous end-joining (NHEJ). While 53BP1 is essential for immunoglobulin heavy chain class switch recombination (CSR), long-range V(D)J recombination and repair of RAG-induced DSBs in XLF-deficient cells, the function of SHLD during these processes remains elusive. Here we report that SHLD1 is dispensable for lymphocyte development and RAG-mediated V(D)J recombination, even in the absence of XLF. By contrast, SHLD1 is essential for restricting resection at AID-induced DSB ends in both NHEJ-proficient and NHEJ-deficient B cells, providing an end-protection mechanism that permits productive CSR by NHEJ and alternative end-joining. Finally, we show that this SHLD1 function is required for orientation-specific joining of AID-initiated DSBs. Our data thus suggest that 53BP1 promotes V(D)J recombination and CSR through two distinct mechanisms: SHLD-independent synapsis of V(D)J segments and switch regions within chromatin, and SHLD-dependent protection of AID-DSB ends against resection.
Sigma-Aldrich
Benzonase® Nuclease, ultrapure, ≥250 units/μL, ≥99% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution, ultrapure grade
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid
Sigma-Aldrich
ATM Kinase Inhibitor, InSolution, ≥95%