MilliporeSigma
  • A First-in-Class, Highly Selective and Cell-Active Allosteric Inhibitor of Protein Arginine Methyltransferase 6.
A First-in-Class, Highly Selective and Cell-Active Allosteric Inhibitor of Protein Arginine Methyltransferase 6.
Journal of medicinal chemistry (2021-02-17)
Yudao Shen, Fengling Li, Magdalena M Szewczyk, Levon Halabelian, Irene Chau, Mohammad S Eram, Carlo Dela Seña, Kwang-Su Park, Fanye Meng, He Chen, Hong Zeng, Aiping Dong, Hong Wu, Viacheslav V Trush, David McLeod, Carlos A Zepeda-Velázquez, Robert M Campbell, Mary M Mader, Brian M Watson, Matthieu Schapira, Cheryl H Arrowsmith, Rima Al-Awar, Dalia Barsyte-Lovejoy, H Ümit Kaniskan, Peter J Brown, Masoud Vedadi, Jian Jin
ABSTRACT
Protein arginine methyltransferase 6 (PRMT6) catalyzes monomethylation and asymmetric dimethylation of arginine residues in various proteins, plays important roles in biological processes, and is associated with multiple cancers. To date, a highly selective PRMT6 inhibitor has not been reported. Here we report the discovery and characterization of a first-in-class, highly selective allosteric inhibitor of PRMT6, (R)-2 (SGC6870). (R)-2 is a potent PRMT6 inhibitor (IC50 = 77 ± 6 nM) with outstanding selectivity for PRMT6 over a broad panel of other methyltransferases and nonepigenetic targets. Notably, the crystal structure of the PRMT6-(R)-2 complex and kinetic studies revealed (R)-2 binds a unique, induced allosteric pocket. Additionally, (R)-2 engages PRMT6 and potently inhibits its methyltransferase activity in cells. Moreover, (R)-2's enantiomer, (S)-2 (SGC6870N), is inactive against PRMT6 and can be utilized as a negative control. Collectively, (R)-2 is a well-characterized PRMT6 chemical probe and a valuable tool for further investigating PRMT6 functions in health and disease.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-dimethyl-Histone H3 (Arg2), clone 20.2 Antibody, rabbit monoclonal, culture supernatant, clone 20.2, Upstate®