MilliporeSigma
  • Effect of Dietary Supplementation with Lipids of Different Unsaturation Degree on Feed Efficiency and Milk Fatty Acid Profile in Dairy Sheep.
Effect of Dietary Supplementation with Lipids of Different Unsaturation Degree on Feed Efficiency and Milk Fatty Acid Profile in Dairy Sheep.
Animals : an open access journal from MDPI (2021-08-28)
Gonzalo Hervás, Pablo G Toral, Cristina Fernández-Díez, Antonella Della Badia, Pilar Frutos
ABSTRACT
Lipids of different unsaturation degree were added to dairy ewe diet to test the hypothesis that unsaturated oils would modulate milk fatty acid (FA) profile without impairing or even improving feed efficiency. To this aim, we examined milk FA profile and efficiency metrics (feed conversion ratio (FCR), energy conversion ratio (ECR), residual feed intake (RFI), and residual energy intake (REI)) in 40 lactating ewes fed a diet with no lipid supplementation (Control) or supplemented with 3 fats rich in saturated, monounsaturated and polyunsaturated FA (i.e., purified palmitic acid (PA), olive oil (OO), and soybean oil (SBO)). Compared with PA, addition of OO decreased milk medium-chain saturated FA and improved the concentration of potentially health-promoting FA, such as cis-9 18:1, trans-11 18:1, cis-9 trans-11 CLA, and 4:0, with no impact on feed efficiency metrics. Nevertheless, FA analysis and decreases in FCR and ECR suggested that SBO supplementation would be a better nutritional strategy to further improve milk FA profile and feed efficiency in dairy ewes. The paradox of differences observed depending on the metric used to estimate feed efficiency (i.e., the lack of variation in RFI and REI vs. changes in FCR and ECR) does not allow solid conclusions to be drawn in this regard.
Sigma-Aldrich
Linoleic acid, conjugated methyl ester
Sigma-Aldrich
Linoleic acid methyl ester, cis/trans-isomers