MilliporeSigma
  • Kupffer cell receptor CLEC4F is important for the destruction of desialylated platelets in mice.
Kupffer cell receptor CLEC4F is important for the destruction of desialylated platelets in mice.
Cell death and differentiation (2021-05-17)
Yizhi Jiang, Yaqiong Tang, Christopher Hoover, Yuji Kondo, Dongping Huang, Damien Restagno, Bojing Shao, Liang Gao, J Michael McDaniel, Meixiang Zhou, Robert Silasi-Mansat, Samuel McGee, Miao Jiang, Xia Bai, Florea Lupu, Changgeng Ruan, Jamey D Marth, Depei Wu, Yue Han, Lijun Xia
ABSTRACT
The liver has recently been identified as a major organ for destruction of desialylated platelets. However, the underlying mechanism remains unclear. Kupffer cells, which are professional phagocytic cells in the liver, comprise the largest population of resident tissue macrophages in the body. Kupffer cells express a C-type lectin receptor, CLEC4F, that recognizes desialylated glycans with an unclear in vivo role in mediating platelet destruction. In this study, we generated a CLEC4F-deficient mouse model (Clec4f-/-) and found that CLEC4F was specifically expressed by Kupffer cells. Using the Clec4f-/- mice and a newly generated platelet-specific reporter mouse line, we revealed a critical role for CLEC4F on Kupffer cells in mediating destruction of desialylated platelets in the liver in vivo. Platelet clearance experiments and ultrastructural analysis revealed that desialylated platelets were phagocytized predominantly by Kupffer cells in a CLEC4F-dependent manner in mice. Collectively, these findings identify CLEC4F as a Kupffer cell receptor important for the destruction of desialylated platelets induced by bacteria-derived neuraminidases, which provide new insights into the pathogenesis of thrombocytopenia in disease conditions such as sepsis.
Product Description
Roche
Neuraminidase (Sialidase), from Arthrobacter ureafaciens