MilliporeSigma
  • No evidence for change in expression of TBC1D1 and TBC1D4 genes in cultured human adipocytes stimulated by myokines and adipokines.
No evidence for change in expression of TBC1D1 and TBC1D4 genes in cultured human adipocytes stimulated by myokines and adipokines.
Adipocyte (2021-03-27)
Łukasz Kępczyński, Szymon Wcisło, Irmina Korzeniewska-Dyl, Katarzyna Połatyńska, Agnieszka Gach, Dariusz Moczulski
ABSTRACT
TBC1D1 and TBC1D4 proteins play analogous, but not identical role in governing insulin-signalling pathway. Little is known about changes in expression levels of TBC1D1 and TBC1D4 genes in mammals, including humans. Number of factors were studied, but data remain controversial. The aim of this study was to evaluate the effect of selected cytokines, adipokines and myokines with known or putative insulin sensitivity regulation activity (adiponectin, irisin, omentin, interleukin 6, leptin, resistin, and tumour necrosis factor) on TBC1D1 and TBC1D4 expression levels in cultured differentiated human adipocytes. No significant differences were found between the adipocytes treated with different stimuli and this effect was determined not dose dependent. It is reasonable to conclude that relative shortage of data showing no change in TBC1D1 and TBC1D4 from literature results from publication bias; therefore, our finding provides additional insight into the role of both genes.
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)
Sigma-Aldrich
DNase I, Amplification Grade