MilliporeSigma
  • Enzymatic activation of pyruvate kinase increases cytosolic oxaloacetate to inhibit the Warburg effect.
Enzymatic activation of pyruvate kinase increases cytosolic oxaloacetate to inhibit the Warburg effect.
Nature metabolism (2021-07-07)
Elizabeth K Wiese, Sadae Hitosugi, Sharon T Loa, Annapoorna Sreedhar, Lindsey G Andres-Beck, Kiran Kurmi, Yuan-Ping Pang, Larry M Karnitz, Wilson I Gonsalves, Taro Hitosugi
ABSTRACT
Pharmacological activation of the glycolytic enzyme PKM2 or expression of the constitutively active PKM1 isoform in cancer cells results in decreased lactate production, a phenomenon known as the PKM2 paradox in the Warburg effect. Here we show that oxaloacetate (OAA) is a competitive inhibitor of human lactate dehydrogenase A (LDHA) and that elevated PKM2 activity increases de novo synthesis of OAA through glutaminolysis, thereby inhibiting LDHA in cancer cells. We also show that replacement of human LDHA with rabbit LDHA, which is relatively resistant to OAA inhibition, eliminated the paradoxical correlation between the elevated PKM2 activity and the decreased lactate concentration in cancer cells treated with a PKM2 activator. Furthermore, rabbit LDHA-expressing tumours, compared to human LDHA-expressing tumours in mice, displayed resistance to the PKM2 activator. These findings describe a mechanistic explanation for the PKM2 paradox by showing that OAA accumulates and inhibits LDHA following PKM2 activation.
Sigma-Aldrich
Anti-PHGDH antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
PKM2 Activator II, DASA, The PKM2 Activator II, DASA modulates the biological activity of PKM2. This small molecule/inhibitor is primarily used for Biochemicals applications.