MilliporeSigma
  • The Contribution of TSLP Activation to Hyperalgesia in Dorsal Root Ganglia Neurons of a Rat.
The Contribution of TSLP Activation to Hyperalgesia in Dorsal Root Ganglia Neurons of a Rat.
International journal of molecular sciences (2022-02-27)
Chun-Ching Lu, Ying-Yi Lu, Hung-Pei Tsai, Chieh-Hsin Wu
ABSTRACT
Peripheral nerve injury involves divergent alterations within dorsal root ganglia (DRG) neurons sensitized by persistent inflammation. Thymic stromal lymphopoietin (TSLP) production is crucial in the development of chronic inflammatory responses. Herein, we investigate the changes of TSLP expression in rats' DRG neurons between injured and uninjured sides in the same rat. Linalyl acetate (LA) was served as a TSLP inhibitor and given intraperitoneally. Rats were assigned to be group of chronic constriction injury (CCI) of the sciatic nerve and the group of CCI of the sciatic nerve administrated with LA. Over 14 days, the rats were measured for paw withdrawal thresholds. DRGs were collected to assess morphological changes via immunofluorescence study. After receiving CCI, the rats rapidly developed mechanical hyperalgesia. TSLP expression at DRG, on the ipsilateral injured side, was consistent with changes in pain behaviors. TSLP appeared in nerve fibers with both small diameters and large diameters. Additionally, TSLP was expressed mostly in transient receptor potential vanilloid-1 (TRPV1)-positive nociceptive neurons. Administration with LA can attenuate the pain behaviors and expression of TSLP in DRG neurons, and in apoptotic neurons at the injured side, but not in the contra-lateral uninjured side. Overall, these results imply that altered expressions of TSLP in nociceptive DRG neurons contributed to mechanical hyperalgesia in a CCI rat model.
Sigma-Aldrich
Anti-TSLP (ab3) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Peripherin antibody, Mouse monoclonal, clone 8G2, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Neurofilament 200 antibody produced in mouse, clone NE14, ascites fluid
Sigma-Aldrich
Anti-Actin Antibody, clone C4, clone C4, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal Anti-CRLF2 antibody produced in mouse, clone 4A11, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse