MilliporeSigma
  • Validation of a deep learning-based image analysis system to diagnose subclinical endometritis in dairy cows.
Validation of a deep learning-based image analysis system to diagnose subclinical endometritis in dairy cows.
PloS one (2022-01-29)
Hafez Sadeghi, Hannah-Sophie Braun, Berner Panti, Geert Opsomer, Osvaldo Bogado Pascottini
ABSTRACT
The assessment of polymorphonuclear leukocyte (PMN) proportions (%) of endometrial samples is the hallmark for subclinical endometritis (SCE) diagnosis. Yet, a non-biased, automated diagnostic method for assessing PMN% in endometrial cytology slides has not been validated so far. We aimed to validate a computer vision software based on deep machine learning to quantify the PMN% in endometrial cytology slides. Uterine cytobrush samples were collected from 116 postpartum Holstein cows. After sampling, each cytobrush was rolled onto three different slides. One slide was stained using Diff-Quick, while a second was stained using Naphthol (golden standard to stain PMN). One single observer evaluated the slides twice at different days under light microscopy. The last slide was stained with a fluorescent dye, and the PMN% were assessed twice by using a fluorescence microscope connected to a smartphone. Fluorescent images were analyzed via the Oculyze Monitoring Uterine Health (MUH) system, which uses a deep learning-based algorithm to identify PMN. Substantial intra-method repeatabilities (via Spearman correlation) were found for Diff-Quick, Naphthol, and Oculyze MUH (r = 0.67 to 0.76). The intra-method agreements (via Kappa value) at ≥1% PMN (κ = 0.44 to 0.47) were lower than at >5 (κ = 0.69 to 0.78) or >10% (κ = 0.67 to 0.85) PMN cut-offs. The inter-method repeatabilities (via Lin's correlation) were also substantial, and values between Diff-Quick and Oculyze MUH, Naphthol and Diff-Quick, and Naphthol and Oculyze MUH were 0.68, 0.69, and 0.77, respectively. The agreements among evaluation methods at ≥1% PMN were weak (κ = 0.06 to 0.28), while it increased at >5 (κ = 0.48 to 0.81) or >10% (κ = 0.50 to 0.65) PMN cut-offs. To conclude, deep learning-based algorithms in endometrial cytology are reliable and useful for simplifying and reducing the diagnosis bias of SCE in dairy cows.
Sigma-Aldrich
Naphthol AS-D chloroacetate, esterase substrate